Heptafluorobutyric anhydride, 98%, Thermo Scientific Chemicals
Heptafluorobutyric anhydride, 98%, Thermo Scientific Chemicals
Heptafluorobutyric anhydride, 98%, Thermo Scientific Chemicals
Heptafluorobutyric anhydride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Heptafluorobutyric anhydride, 98%, Thermo Scientific Chemicals
CAS: 336-59-4 | C8F14O3 | 410.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR169260050
Price (MYR)
732.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
732.00
Add to cart
Specifications
Chemical Name or MaterialHeptafluorobutyric anhydride
CAS336-59-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
View more
Acylation reagent
RUO – Research Use Only