4-tert-Butylbenzoyl chloride, 98%, Thermo Scientific Chemicals
4-tert-Butylbenzoyl chloride, 98%, Thermo Scientific Chemicals
4-tert-Butylbenzoyl chloride, 98%, Thermo Scientific Chemicals
4-tert-Butylbenzoyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-tert-Butylbenzoyl chloride, 98%, Thermo Scientific Chemicals
CAS: 1710-98-1 | C11H13ClO | 196.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR169100250
Price (MYR)
829.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
829.00
Add to cart
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size-20+40 mesh (425-850 micron)