Cyclohexanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals
Cyclohexanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals
Cyclohexanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals
Cyclohexanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals
CAS: 2719-27-9 | C7H11ClO | 146.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR169050250
Price (MYR)
469.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
469.00
Add to cart
Specifications
Chemical Name or MaterialCyclohexanecarboxylic acid chloride
Name Note98%
CAS2719-27-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement: