2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 99+%, Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 99+%, Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 99+%, Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 99+%, Thermo Scientific Chemicals
CAS: 6976-37-0 | C8H19NO5 | 209.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number ACR168871000
Price (MYR)
800.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
800.00
Add to cart
Specifications
Chemical Name or Material2,2-Bis(hydroxymethyl)-2,2′,2′′-nitrilotriethanol
Name Note99+%
CAS6976-37-0
Absorbance(0.1M in H2O)
CAS Min %99.0