Phenanthrenequinone, 95%, Thermo Scientific Chemicals
Phenanthrenequinone, 95%, Thermo Scientific Chemicals
Phenanthrenequinone, 95%, Thermo Scientific Chemicals
Phenanthrenequinone, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenanthrenequinone, 95%, Thermo Scientific Chemicals
CAS: 84-11-7 | C14H8O2 | 208.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR168790250
Price (MYR)
628.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
628.00
Add to cart
Chemical Identifiers
CAS76347-13-2
IUPAC Name4,4,5,5-tetramethyl-2-(propan-2-yl)-1,3,2-dioxaborolane
Molecular FormulaC9H19BO2
InChI KeyMECCSFDHAVAAAW-UHFFFAOYSA-N
SMILESCC(C)B1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4070-1.4110 @ 20°C
Appearance (Color)Clear colorless
Assay (GC)≥96.0%