6-Chloronicotinic acid, 99%, Thermo Scientific Chemicals
6-Chloronicotinic acid, 99%, Thermo Scientific Chemicals
6-Chloronicotinic acid, 99%, Thermo Scientific Chemicals
6-Chloronicotinic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
6-Chloronicotinic acid, 99%, Thermo Scientific Chemicals
CAS: 5326-23-8 | C6H4ClNO2 | 157.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number ACR168690250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2869-83-2
IUPAC Name3-(diethylamino)-7-{2-[4-(dimethylamino)phenyl]diazen-1-yl}-5-phenyl-5λ⁵-phenazin-5-ylium chloride
Molecular FormulaC30H31ClN6
InChI KeyXXACTDWGHQXLGW-UHFFFAOYSA-M
SMILES[Cl-].CCN(CC)C1=CC=C2N=C3C=CC(=CC3=[N+](C3=CC=CC=C3)C2=C1)N=NC1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green to dark brown to black
Infrared spectrumConforms
Appearance (Form)Powder
Dye content>=50 %