Oxonic acid, potassium salt, 97.5%
Oxonic acid, potassium salt, 97.5%
Oxonic acid, potassium salt, 97.5%
Oxonic acid, potassium salt, 97.5%
Thermo Scientific Chemicals
Oxonic acid, potassium salt, 97.5%
CAS: 2207-75-2 | C4H2KN3O4 | 195.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR168530250
Price (MYR)
1,186.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,186.00
Add to cart
Specifications
Chemical Name or MaterialOxonic acid, potassium salt
Name Note97.5%
CAS2207-75-2
Melting Point>300.0°C