2,2,4-Trimethylpentane, 99%, for spectroscopy, Thermo Scientific Chemicals
CAS: 540-84-1 | C8H18 | 114.232 g/mol
Have Questions?
Change view
Quantity:
2.5 L
500 mL
Packaging:
Glass Bottle
Catalog number ACR167910025
Price (MYR)
1,189.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,189.00
Add to cart
Specifications
Chemical Name or Material2, 2, 4-Trimethylpentane
Melting Point-107.0°C
Name Note99%
Absorbance0.01 max. at 250nm, 0.01 max. at 400nm, 0.04 max. at 240nm, 0.1 max. at 230nm, 0.2 max. at 220nm, 1 max. at 210nm
CAS540-84-1