n-heptane, for HPLC, ∽ 95% n-heptane, Thermo Scientific Chemicals
n-heptane, for HPLC, ∽ 95% n-heptane, Thermo Scientific Chemicals
n-heptane, for HPLC, ∽ 95% n-heptane, Thermo Scientific Chemicals
n-heptane, for HPLC, ∽ 95% n-heptane, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-heptane, for HPLC, ∽ 95% n-heptane, Thermo Scientific Chemicals
CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number ACR167820010
Price (MYR)
569.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
569.00
Add to cart
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA99.9% (metals basis)
Appearance (Color)Silvery white
FormWire, 0.404mm (0.0159in) diameter, annealed.
For HPLC
RUO – Research Use Only