Decanoic acid, 99%, Thermo Scientific Chemicals
Decanoic acid, 99%, Thermo Scientific Chemicals
Decanoic acid, 99%, Thermo Scientific Chemicals
Decanoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Decanoic acid, 99%, Thermo Scientific Chemicals
CAS: 334-48-5 | C10H20O2 | 172.268 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
Catalog number ACR167271000
Price (MYR)
482.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
482.00
Add to cart
Specifications
Chemical Name or MaterialDecanoic acid
Name Note99+%
CAS334-48-5
Melting Point30.0°C to 32.0°C
Boiling Point268.0°C to 270.0°C