Decanoic acid, 99%, Thermo Scientific Chemicals
Decanoic acid, 99%, Thermo Scientific Chemicals
Decanoic acid, 99%, Thermo Scientific Chemicals
Decanoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Decanoic acid, 99%, Thermo Scientific Chemicals
CAS: 334-48-5 | C10H20O2 | 172.268 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR167270010
Price (MYR)
1,603.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,603.00
Add to cart
Chemical Identifiers
CAS3685-23-2
IUPAC Name4-aminocyclohexane-1-carboxylic acid
Molecular FormulaC7H13NO2
InChI KeyDRNGLYHKYPNTEA-UHFFFAOYSA-N
SMILESNC1CCC(CC1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Non-aqueous acid-base Titration)≥96.0% to ≤104.0%
Appearance (Color)White