4-Chlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals
4-Chlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals
4-Chlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals
4-Chlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals
CAS: 122-88-3 | C8H7ClO3 | 186.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number ACR166965000
Price (MYR)
1,063.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,063.00
Add to cart
Chemical Identifiers
CAS78-90-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Refractive index1.4450 to 1.4470 (20°C, 589 nm)
Color scale=<20 APHA
GC>=98.5 %
Water=<0.5 %