4-Chlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals
4-Chlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals
4-Chlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals
4-Chlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chlorophenoxyacetic acid, 98%, Thermo Scientific Chemicals
CAS: 122-88-3 | C8H7ClO3 | 186.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number ACR166961000
Price (MYR)
464.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
464.00
Add to cart
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
CommentNo Specifications