5-Bromo-2-thiophenecarboxaldehyde, 97%, Thermo Scientific Chemicals
5-Bromo-2-thiophenecarboxaldehyde, 97%, Thermo Scientific Chemicals
5-Bromo-2-thiophenecarboxaldehyde, 97%, Thermo Scientific Chemicals
5-Bromo-2-thiophenecarboxaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Bromo-2-thiophenecarboxaldehyde, 97%, Thermo Scientific Chemicals
CAS: 4701-17-1 | C5H3BrOS | 191.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
Packaging:
Glass bottle
Catalog number ACR166650050
Price (MYR)
566.00
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
566.00
Add to cart
Chemical Identifiers
CAS128-08-5
IUPAC Name1-bromopyrrolidine-2,5-dione
Molecular FormulaC4H4BrNO2
InChI KeyPCLIMKBDDGJMGD-UHFFFAOYSA-N
SMILESBrN1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
Loss on drying=<1 %
Appearance (Form)Crystalline powder
Titration Iodometric98.5 to 101.5 % (on dry substance)