1-(2-Pyridyl)piperazine, 98%, Thermo Scientific Chemicals
1-(2-Pyridyl)piperazine, 98%, Thermo Scientific Chemicals
1-(2-Pyridyl)piperazine, 98%, Thermo Scientific Chemicals
1-(2-Pyridyl)piperazine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-(2-Pyridyl)piperazine, 98%, Thermo Scientific Chemicals
CAS: 34803-66-2 | C9H13N3 | 163.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR165900250
Price (MYR)
1,074.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,074.00
Add to cart
Chemical Identifiers
CAS206752-32-1
IUPAC Namesodium 4-hydroxy-5-{[(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}-7-sulfonaphthalene-2-sulfonate
Molecular FormulaC17H12NNaO8S2
InChI KeyGRYBZNLNHVSHPY-UHFFFAOYSA-M
SMILES[Na+].OC1=C2C(NC=C3C=CC=CC3=O)=CC(=CC2=CC(=C1)S([O-])(=O)=O)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
Assay (ex Sulphated Ash)> 85.0% (with reference to dried material)
Water Content (Karl Fischer Titration)≤ 8.0%