Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Glass bottle
Catalog number ACR165895000
Price (MYR)
885.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
885.00
Add to cart
Chemical Identifiers
CAS2498-50-2
IUPAC Namedihydrogen 4-aminobenzene-1-carboximidamide dichloride
Molecular FormulaC7H11Cl2N3
InChI KeyGHEHNICLPWTXJC-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].NC(=N)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Appearance (Color)White to yellow
Titration Argentometric>=96.0 %