Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number ACR165891000
Price (MYR)
451.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
451.00
Add to cart
Specifications
Chemical Name or MaterialGlycidyl methacrylate
CAS106-91-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic in contact with skin.
Suspected of causing genetic defects.
May damage fertility.
May cau
CAS150-76-5