Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycidyl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR165890025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS501-30-4
IUPAC Name5-hydroxy-2-(hydroxymethyl)-4H-pyran-4-one
Molecular FormulaC6H6O4
InChI KeyBEJNERDRQOWKJM-UHFFFAOYSA-N
SMILESOCC1=CC(=O)C(O)=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point151°C to 156°C
Infrared spectrumConforms
Appearance (Color)White to beige to light brown
Titration with NaOH>=98.5 %