Isoamyl nitrite, 97%, stabilized, Thermo Scientific Chemicals
Isoamyl nitrite, 97%, stabilized, Thermo Scientific Chemicals
Isoamyl nitrite, 97%, stabilized, Thermo Scientific Chemicals
Isoamyl nitrite, 97%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isoamyl nitrite, 97%, stabilized, Thermo Scientific Chemicals
CAS: 110-46-3 | C5H11NO2 | 117.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR165705000500 mLGlass Bottle
Catalog number ACR165705000
Price (MYR)
1,187.00
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,187.00
Add to cart
Chemical Identifiers
CAS68-12-2
IUPAC NameN,N-dimethylformamide
Molecular FormulaC3H7NO
InChI KeyZMXDDKWLCZADIW-UHFFFAOYSA-N
SMILESCN(C)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.5 %
Acidity/alkalinity=<0.0005 meq/g
Calcium (Ca)=<1 ppm
Iron (Fe)=<1 ppm
Magnesium (Mg)=<0.5 ppm