Phenyl salicylate, 99%, Thermo Scientific Chemicals
Phenyl salicylate, 99%, Thermo Scientific Chemicals
Phenyl salicylate, 99%, Thermo Scientific Chemicals
Phenyl salicylate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyl salicylate, 99%, Thermo Scientific Chemicals
Phenyl salicylate, 99%, C13H10O3, CAS Number-118-55-8, salol, salphenyl, phenyl salicylate, 2-phenoxycarbonylphenol, salicylic acid, phenyl ester, benzoic acid, 2-hydroxy-, phenyl ester, phenyl-2-hydroxybenzoate, musol, phenol salicylate, 2-hydroxybenzoic acid phenyl ester, 250g, 98.5% min. (GC)
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number ACR165082500
Price (MYR)
493.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
493.00
Add to cart
Chemical Identifiers
CAS75-30-9
IUPAC Name2-iodopropane
Molecular FormulaC3H7I
InChI KeyFMKOJHQHASLBPH-UHFFFAOYSA-N
SMILESCC(C)I
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4960-1.5000 @ 20?C
Assay (GC)≥98.0%
Identification (FTIR)Conforms
StabilizerCopper powder