Phenyl salicylate, 99%, Thermo Scientific Chemicals
Phenyl salicylate, 99%, Thermo Scientific Chemicals
Phenyl salicylate, 99%, Thermo Scientific Chemicals
Phenyl salicylate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyl salicylate, 99%, Thermo Scientific Chemicals
Phenyl salicylate, 99%, C13H10O3, CAS Number-118-55-8, salol, salphenyl, phenyl salicylate, 2-phenoxycarbonylphenol, salicylic acid, phenyl ester, benzoic acid, 2-hydroxy-, phenyl ester, phenyl-2-hydroxybenzoate, musol, phenol salicylate, 2-hydroxybenzoic acid phenyl ester, 250g, 98.5% min. (GC)
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Glass bottle
Catalog number ACR165080010
Price (MYR)
1,200.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,200.00
Add to cart
Chemical Identifiers
CAS5653-40-7
IUPAC Name2-amino-4,5-dimethoxybenzoic acid
Molecular FormulaC9H11NO4
InChI KeyHJVAVGOPTDJYOJ-UHFFFAOYSA-N
SMILESCOC1=CC(N)=C(C=C1OC)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to dark brown to brown or purple to pale gray to gray
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Refractive Index1.5030-1.5070 @ 20?C
FormPowder or clear liquid as melt