2-Iodopropane, 98+%, stabilized, Thermo Scientific Chemicals
2-Iodopropane, 98+%, stabilized, Thermo Scientific Chemicals
2-Iodopropane, 98+%, stabilized, Thermo Scientific Chemicals
2-Iodopropane, 98+%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Iodopropane, 98+%, stabilized, Thermo Scientific Chemicals
CAS: 75-30-9 | C3H7I | 169.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number ACR164965000
Price (MYR)
2,654.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,654.00
Add to cart
Chemical Identifiers
CAS126213-50-1
IUPAC Name2H,3H-thieno[3,4-b][1,4]dioxine
Molecular FormulaC6H6O2S
InChI KeyGKWLILHTTGWKLQ-UHFFFAOYSA-N
SMILESC1COC2=CSC=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥96.0% (GC)
FormLiquid
Appearance (Color)Colorless to yellow