4,4'-Bis(dimethylamino)benzophenone, 98%, Thermo Scientific Chemicals
4,4'-Bis(dimethylamino)benzophenone, 98%, Thermo Scientific Chemicals
4,4'-Bis(dimethylamino)benzophenone, 98%, Thermo Scientific Chemicals
4,4'-Bis(dimethylamino)benzophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4,4'-Bis(dimethylamino)benzophenone, 98%, Thermo Scientific Chemicals
CAS: 90-94-8 | C17H20N2O | 268.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR164490050
Price (MYR)
380.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
380.00
Add to cart
Chemical Identifiers
CAS368-90-1
IUPAC Name[4-(trifluoromethyl)phenyl]hydrazine
Molecular FormulaC7H7F3N2
InChI KeyDBNLGTYGKCMLLR-UHFFFAOYSA-N
SMILESNNC1=CC=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Identification (FTIR)Conforms
Appearance (Color)Pale yellow to orange or pale brown
Assay (GC)≥94.0%
Melting Point (clear melt)58-65?C