2-Hydroxy-5-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
2-Hydroxy-5-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
2-Hydroxy-5-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
2-Hydroxy-5-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Hydroxy-5-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
CAS: 672-13-9 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
5 g
Packaging:
Glass bottle
Catalog number ACR164160100
Price (MYR)
1,039.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,039.00
Add to cart
Chemical Identifiers
CAS1344-28-1
IUPAC Namedialuminium(3+) trioxidandiide
Molecular FormulaAl2O3
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Sodium oxide (Na2O)=<0.4 %
Silicon dioxide (SiO2)=<0.03 %
Appearance (Color)White to off-white
pH6.5 to 7.5 (10% aqueous suspension)
Particle size=<5 % (<40 µm)