2-Hydroxy-5-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
2-Hydroxy-5-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
2-Hydroxy-5-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
2-Hydroxy-5-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Hydroxy-5-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
CAS: 672-13-9 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
Packaging:
Glass bottle
Catalog number ACR164160050
Price (MYR)
732.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
732.00
Add to cart
Chemical Identifiers
CAS7758-99-8
IUPAC Namecopper(2+) pentahydrate sulfate
Molecular FormulaCuH10O9S
InChI KeyJZCCFEFSEZPSOG-UHFFFAOYSA-L
SMILESO.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue
FormCrystals or crystalline powder and/or lumps
Assay (unspecified)>98.5 to <101.5%