Benzyltriethylammonium chloride, 98%, Thermo Scientific Chemicals
Benzyltriethylammonium chloride, 98%, Thermo Scientific Chemicals
Benzyltriethylammonium chloride, 98%, Thermo Scientific Chemicals
Benzyltriethylammonium chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzyltriethylammonium chloride, 98%, Thermo Scientific Chemicals
CAS: 56-37-1 | C13H22ClN | 227.78 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number ACR164021000
Price (MYR)
394.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
394.00
Add to cart
Chemical Identifiers
CAS22726-00-7
IUPAC Name3-bromobenzamide
Molecular FormulaC7H6BrNO
InChI KeyODJFDWIECLJWSR-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.0%
Appearance (Color)White to off-white
FormPowder or crystalline powder
Phase transfer catalyst
RUO – Research Use Only