2-Methyl-4-nitroaniline, 99%, Thermo Scientific Chemicals
2-Methyl-4-nitroaniline, 99%, Thermo Scientific Chemicals
2-Methyl-4-nitroaniline, 99%, Thermo Scientific Chemicals
2-Methyl-4-nitroaniline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Methyl-4-nitroaniline, 99%, Thermo Scientific Chemicals
CAS: 99-52-5 | C7H8N2O2 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number ACR163950050
Price (MYR)
367.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
367.00
Add to cart
Chemical Identifiers
CAS42454-06-8
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Crystalline powder
Melting point165°C to 170°C
Appearance (Color)Yellow