5-Methoxysalicylic acid, 98%
5-Methoxysalicylic acid, 98%
5-Methoxysalicylic acid, 98%
5-Methoxysalicylic acid, 98%
5-Methoxysalicylic acid, 98%
Thermo Scientific Chemicals
5-Methoxysalicylic acid, 98%
CAS: 2612-02-4 | C8H8O4 | 168.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR16387010010 gGlass bottle
Catalog number ACR163870100
Price (MYR)
472.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
472.00
Add to cart
Chemical Identifiers
CAS629-96-9
IUPAC Nameicosan-1-ol
Molecular FormulaC20H42O
InChI KeyBTFJIXJJCSYFAL-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or lumps or flakes or pellets
Assay (GC)≥95.0%
Melting Point (clear melt)60.0-70.0?C