5-Methylsalicylic acid, 98%
5-Methylsalicylic acid, 98%
5-Methylsalicylic acid, 98%
5-Methylsalicylic acid, 98%
Thermo Scientific Chemicals
5-Methylsalicylic acid, 98%
CAS: 89-56-5 | C8H8O3 | 152.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR16386010010 gGlass bottle
Catalog number ACR163860100
Price (MYR)
588.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
588.00
Add to cart
Chemical Identifiers
CAS15489-27-7
IUPAC Namecopper(2+) dilithium(1+) tetrachloride
Molecular FormulaCl4CuLi2
InChI KeyHCJWWBBBSCXJMS-UHFFFAOYSA-J
SMILES[Li+].[Li+].[Cl-].[Cl-].[Cl-].[Cl-].[Cu++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Clear orange to red-brown
Titration Argentometric0.09 to 0.12 M