2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%, Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%, Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%, Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%, Thermo Scientific Chemicals
CAS: 6976-37-0 | C8H19NO5 | 209.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ACR163825000
Price (MYR)
1,848.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,848.00
Add to cart
Chemical Identifiers
CAS403-42-9
IUPAC Name1-(4-fluorophenyl)ethan-1-one
Molecular FormulaC8H7FO
InChI KeyZDPAWHACYDRYIW-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=98.5 %
Refractive index1.5090 to 1.5120 (20°C, 589 nm)