2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%, Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%, Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%, Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%, Thermo Scientific Chemicals
CAS: 6976-37-0 | C8H19NO5 | 209.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ACR163821000
Price (MYR)
751.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
751.00
Add to cart
Chemical Identifiers
CAS1533-20-6
IUPAC Name1,3-diphenylbutan-1-one
Molecular FormulaC16H16O
InChI KeyGIVFXLVPKFXTCU-UHFFFAOYNA-N
SMILESCC(CC(=O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or flakes
Assay (GC)≥94.0%
Appearance (Color)Pale yellow
Melting Point (clear melt)65.5-74.5?C