Adenine sulfate, 98%, synthetic
Adenine sulfate, 98%, synthetic
Adenine sulfate, 98%, synthetic
Adenine sulfate, 98%, synthetic
Thermo Scientific Chemicals
Adenine sulfate, 98%, synthetic
CAS: 321-30-2 | C10H12N10O4S | 368.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number ACR163630250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS652148-91-9
IUPAC Name(5-chloropyridin-2-yl)boronic acid
Molecular FormulaC5H5BClNO2
InChI KeyJEGHCYRKSUGHJH-UHFFFAOYSA-N
SMILESOB(O)C1=NC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%