Brucine dihydrate, 99%, Thermo Scientific Chemicals
Brucine dihydrate, 99%, Thermo Scientific Chemicals
Brucine dihydrate, 99%, Thermo Scientific Chemicals
Brucine dihydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Brucine dihydrate, 99%, Thermo Scientific Chemicals
CAS: 145428-94-0 | C23H27N2O4 | 395.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number ACR163500500
Price (MYR)
861.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
861.00
Add to cart
Specifications
Chemical Name or MaterialBrucine dihydrate
Name Note99%
CAS145428-94-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Harmful to aquatic life with long lasting effects.
Fatal if swallowed.
Fatal if inhaled.