α-Bromo-p-tolunitrile, 98%, Thermo Scientific Chemicals
α-Bromo-p-tolunitrile, 98%, Thermo Scientific Chemicals
α-Bromo-p-tolunitrile, 98%, Thermo Scientific Chemicals
α-Bromo-p-tolunitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
α-Bromo-p-tolunitrile, 98%, Thermo Scientific Chemicals
CAS: 17201-43-3 | C8H6BrN | 196.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number ACR162760100
Price (MYR)
686.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
686.00
Add to cart
Chemical Identifiers
CAS2386-53-0
IUPAC Namesodium dodecane-1-sulfonate
Molecular FormulaC12H25NaO3S
InChI KeyDAJSVUQLFFJUSX-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=99.0 %
Appearance (Color)White to cream
Appearance (Form)Powder