Diglycolic acid, 98%, Thermo Scientific Chemicals
Diglycolic acid, 98%, Thermo Scientific Chemicals
Diglycolic acid, 98%, Thermo Scientific Chemicals
Diglycolic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diglycolic acid, 98%, Thermo Scientific Chemicals
CAS: 110-99-6 | C4H6O5 | 134.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR162261000
Price (MYR)
634.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
634.00
Add to cart
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystalline powder
Total Metal Impurities≤1.0%