Diglycolic acid, 98%, Thermo Scientific Chemicals
Diglycolic acid, 98%, Thermo Scientific Chemicals
Diglycolic acid, 98%, Thermo Scientific Chemicals
Diglycolic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diglycolic acid, 98%, Thermo Scientific Chemicals
CAS: 110-99-6 | C4H6O5 | 134.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
Packaging:
Plastic bottle
Catalog number ACR162260025
Price (MYR)
7,166.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
7,166.00
Add to cart
Chemical Identifiers
CAS85684-61-3
IUPAC Name3-(difluoromethoxy)benzaldehyde
Molecular FormulaC8H6F2O2
InChI KeyHFIUSWPRDIPIPN-UHFFFAOYSA-N
SMILESFC(F)OC1=CC=CC(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)≤1.5%
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Refractive Index1.4920-1.4970 @ 20?C