N,N-Dimethylformamide diethyl acetal, 95%, Thermo Scientific Chemicals
N,N-Dimethylformamide diethyl acetal, 95%, Thermo Scientific Chemicals
N,N-Dimethylformamide diethyl acetal, 95%, Thermo Scientific Chemicals
N,N-Dimethylformamide diethyl acetal, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylformamide diethyl acetal, 95%, Thermo Scientific Chemicals
CAS: 1188-33-6 | C7H17NO2 | 147.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Catalog number ACR162080500
Price (MYR)
1,531.00
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
1,531.00
Add to cart
Chemical Identifiers
CAS99-33-2
IUPAC Name3,5-dinitrobenzoyl chloride
Molecular FormulaC7H3ClN2O5
InChI KeyNNOHXABAQAGKRZ-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(=CC(=C1)C(Cl)=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to beige
Titration RCOCl>=98.5 %
Appearance (Form)Crystals or crystalline needles
Infrared spectrumConforms
Melting point67°C to 71°C
Alkylation reagent
RUO – Research Use Only