Phenyl benzoate, 99%, Thermo Scientific Chemicals
Phenyl benzoate, 99%, Thermo Scientific Chemicals
Phenyl benzoate, 99%, Thermo Scientific Chemicals
Phenyl benzoate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyl benzoate, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number ACR162061000
Price (MYR)
550.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
550.00
Add to cart
Specifications
Chemical Name or MaterialPhenyl benzoate
Name Note99%
Acidity1mg KOH/g max.
CAS93-99-2
Health Hazard 1GHS Signal Word: Warning