3-Chloropropylamine hydrochloride, 98%, Thermo Scientific Chemicals
3-Chloropropylamine hydrochloride, 98%, Thermo Scientific Chemicals
3-Chloropropylamine hydrochloride, 98%, Thermo Scientific Chemicals
3-Chloropropylamine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Chloropropylamine hydrochloride, 98%, Thermo Scientific Chemicals
CAS: 6276-54-6 | C3H8ClN·HCl | 130.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Plastic bottle
Catalog number ACR162010500
Price (MYR)
415.00
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
415.00
Add to cart
Specifications
Chemical Name or Material3-Chloropropylamine hydrochloride
Name Note98%
CAS6276-54-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning