2,4,6-Collidine, 99%, Thermo Scientific Chemicals
2,4,6-Collidine, 99%, Thermo Scientific Chemicals
2,4,6-Collidine, 99%, Thermo Scientific Chemicals
2,4,6-Collidine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,4,6-Collidine, 99%, Thermo Scientific Chemicals
CAS: 108-75-8 | C8H11N | 121.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ACR161955000
Price (MYR)
1,620.00
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
1,620.00
Add to cart
Specifications
Chemical Name or Material2, 4, 6-Collidine
CAS108-75-8
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0