2,4,6-Collidine, 99%, Thermo Scientific Chemicals
2,4,6-Collidine, 99%, Thermo Scientific Chemicals
2,4,6-Collidine, 99%, Thermo Scientific Chemicals
2,4,6-Collidine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,4,6-Collidine, 99%, Thermo Scientific Chemicals
CAS: 108-75-8 | C8H11N | 121.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number ACR161951000
Price (MYR)
716.00
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
716.00
Add to cart
Chemical Identifiers
CAS7558-79-4
IUPAC Namedisodium hydrogen phosphate
Molecular FormulaHNa2O4P
InChI KeyBNIILDVGGAEEIG-UHFFFAOYSA-L
SMILES[Na+].[Na+].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Titration with HClO498.0 to 102.0 % (On dried substance)
Loss on drying=<5 % (130°C)
pH8.7 to 9.4 (5 % in water at 25°C)