2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline, 98%, Thermo Scientific Chemicals
2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline, 98%, Thermo Scientific Chemicals
2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline, 98%, Thermo Scientific Chemicals
2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline, 98%, Thermo Scientific Chemicals
CAS: 4733-39-5 | C26H20N2 | 360.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR161340050
Price (MYR)
2,361.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,361.00
Add to cart
Chemical Identifiers
CAS609-73-4
IUPAC Name1-iodo-2-nitrobenzene
Molecular FormulaC6H4INO2
InChI KeyJXMZUNPWVXQADG-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC=C1I
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder mixed with darker coarse granules and/or lumps
Melting Point (clear melt)48-53?C
Identification (FTIR)Conforms
Assay (GC)≥96.0%
Appearance (Color)Yellow to brown