2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline, 98%, Thermo Scientific Chemicals
2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline, 98%, Thermo Scientific Chemicals
2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline, 98%, Thermo Scientific Chemicals
2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline, 98%, Thermo Scientific Chemicals
CAS: 4733-39-5 | C26H20N2 | 360.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR161340010
Price (MYR)
772.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
772.00
Add to cart
Chemical Identifiers
CAS79-09-4
IUPAC Namepropanoic acid
Molecular FormulaC3H6O2
InChI KeyXBDQKXXYIPTUBI-UHFFFAOYSA-N
SMILESCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Water=<0.1 %
Appearance (Form)Clear liquid
Infrared spectrumConforms
Refractive index1.3850 to 1.3870 (20°C, 589 nm)