N,N-Dimethylformamide dimethyl acetal, ca. 97%, Thermo Scientific Chemicals
N,N-Dimethylformamide dimethyl acetal, ca. 97%, Thermo Scientific Chemicals
N,N-Dimethylformamide dimethyl acetal, ca. 97%, Thermo Scientific Chemicals
N,N-Dimethylformamide dimethyl acetal, ca. 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylformamide dimethyl acetal, ca. 97%, Thermo Scientific Chemicals
CAS: 4637-24-5 | C5H13NO2 | 119.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR161250250
Price (MYR)
464.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
464.00
Add to cart
Chemical Identifiers
CAS77613-65-1
IUPAC Name2-(bromomethyl)-1,1-difluorocyclopropane
Molecular FormulaC4H5BrF2
InChI KeyTUDJNSKRXIUOAJ-UHFFFAOYNA-N
SMILESFC1(F)CC1CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Refractive Index1.4220-1.4270 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Alkylation reagent
RUO – Research Use Only