3-Methoxyphenol, 97%, Thermo Scientific Chemicals
3-Methoxyphenol, 97%, Thermo Scientific Chemicals
3-Methoxyphenol, 97%, Thermo Scientific Chemicals
3-Methoxyphenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Methoxyphenol, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number ACR161061000
Price (MYR)
847.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
847.00
Add to cart
Specifications
Chemical Name or Material3-Methoxyphenol
CAS150-19-6
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0