Phenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Phenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Phenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Phenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyl isothiocyanate, 98%, Thermo Scientific Chemicals
CAS: 103-72-0 | C7H5NS | 135.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number ACR160975000
Price (MYR)
1,435.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,435.00
Add to cart
Chemical Identifiers
CAS4930-98-7
IUPAC Name2-hydrazinylpyridine
Molecular FormulaC5H7N3
InChI KeyNWELCUKYUCBVKK-UHFFFAOYSA-N
SMILESNNC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals or crystalline mass
Melting point43°C to 47°C
Infrared spectrumConforms
Appearance (Color)White to orange-brown
GC>=97.5 %