Phenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Phenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Phenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Phenyl isothiocyanate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyl isothiocyanate, 98%, Thermo Scientific Chemicals
CAS: 103-72-0 | C7H5NS | 135.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR160971000
Price (MYR)
674.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
674.00
Add to cart
Chemical Identifiers
CAS45644-21-1
IUPAC Name6-chloropyridin-2-amine
Molecular FormulaC5H5ClN2
InChI KeyOBYJTLDIQBWBHM-UHFFFAOYSA-N
SMILESNC1=CC=CC(Cl)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Form)Crystalline powder or solid
Melting point69°C to 75°C
Appearance (Color)White to brown