L(-)-Borneol, 97%, Thermo Scientific Chemicals
L(-)-Borneol, 97%, Thermo Scientific Chemicals
L(-)-Borneol, 97%, Thermo Scientific Chemicals
L(-)-Borneol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(-)-Borneol, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number ACR160651000
Price (MYR)
683.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
683.00
Add to cart
Chemical Identifiers
CAS3710-84-7
IUPAC NameN,N-diethylhydroxylamine
Molecular FormulaC4H11NO
InChI KeyFVCOIAYSJZGECG-UHFFFAOYSA-N
SMILESCCN(O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<100 APHA
GC>=96.0 %
Refractive index1.4190 to 1.4220 (20°C, 589 nm)
Appearance (Form)Clear liquid