L(-)-Borneol, 97%, Thermo Scientific Chemicals
L(-)-Borneol, 97%, Thermo Scientific Chemicals
L(-)-Borneol, 97%, Thermo Scientific Chemicals
L(-)-Borneol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(-)-Borneol, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number ACR160650250
Price (MYR)
491.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
491.00
Add to cart
Chemical Identifiers
CAS59-02-9
IUPAC Name(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol
Molecular FormulaC29H50O2
InChI KeyGVJHHUAWPYXKBD-IEOSBIPESA-N
SMILESCC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Viscous liquid
GC>=97.0 %
Refractive index1.5050 to 1.5070 (20°C, 589 nm)
Appearance (Color)Clear yellow to red-brown