1-Chloro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Chloro-2,4-dinitrobenzene, 99%, Thermo Scientific Chemicals
CAS: 97-00-7 | C6H3ClN2O4 | 202.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR160515000
Price (MYR)
784.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
784.00
Add to cart
Chemical Identifiers
CAS98-95-3
IUPAC Namenitrobenzene
Molecular FormulaC6H5NO2
InChI KeyLQNUZADURLCDLV-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Infrared spectrumConforms
Refractive index1.5520 to 1.5540 (20°C, 589 nm)
Calcium (Ca)=<0.5 ppm
Cadmium (Cd)=<0.05 ppm